Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MMP7
Entrez Gene ID
4316
Gene Name
matrix metallopeptidase 7
Gene Aliases
MMP-7, MPSL1, PUMP-1
Chromosome Location
Chr.11: 102520508 - 102530753 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_002423.4 NP_002414.1 1 - 2 160 101
GenBank mRNA AK222980.1 - 1 - 2 160 101
AK293359.1 - 1 - 2 154 101
AK309687.1 - 1 - 2 157 101
BC003635.2 - 1 - 2 138 101
BP332706.1 - 1 - 2 154 101
X07819.1 - 1 - 2 134 101
Z11887.1 - 1 - 2 134 101

View Full Product Details