The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002430.2 | NP_002421.3 | 1 - 2 | 4738 | 62 |
GenBank mRNA | X82209.2 | - | 1 - 2 | 4736 | 62 |