The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002496.3 | NP_002487.1 | 5 - 6 | 488 | 52 |
XM_005274013.1 | XP_005274070.1 | 5 - 6 | 585 | 52 | |
XM_005274014.2 | XP_005274071.1 | 5 - 6 | 612 | 52 | |
XM_011545053.2 | XP_011543355.1 | 5 - 6 | 523 | 52 | |
GenBank mRNA | AK223114.1 | - | 5 - 6 | 478 | 52 |
AK314546.1 | - | 5 - 6 | 413 | 52 | |
BC119754.2 | - | 5 - 6 | 417 | 52 | |
U65579.1 | - | 5 - 6 | 411 | 52 |