The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001270616.1 | NP_001257545.1 | 2 - 3 | 2342 | 128 |
NM_002763.4 | NP_002754.2 | 2 - 3 | 2015 | 128 | |
XM_005273194.1 | XP_005273251.1 | 2 - 3 | 2341 | 128 | |
XM_005273195.4 | XP_005273252.1 | 2 - 3 | 2370 | 128 | |
XM_011509771.1 | XP_011508073.1 | 3 - 4 | 2449 | 128 | |
XM_011509772.2 | XP_011508074.1 | 2 - 3 | 2238 | 128 | |
XM_011509773.2 | XP_011508075.1 | 2 - 3 | 1879 | 128 | |
XM_017001832.1 | XP_016857321.1 | 2 - 3 | 2199 | 128 | |
XM_017001833.1 | XP_016857322.1 | 2 - 3 | 2199 | 128 | |
GenBank mRNA | AK298182.1 | - | 2 - 3 | 570 | 128 |
BC024201.2 | - | 2 - 3 | 1999 | 128 | |
U44060.1 | - | 2 - 3 | 2330 | 128 |