Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PYGL
Entrez Gene ID
5836
Gene Name
phosphorylase, glycogen, liver
Gene Aliases
GSD6
Chromosome Location
Chr.14: 50905217 - 50944530 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001163940.1 NP_001157412.1 18 - 19 2410 97
NM_002863.4 NP_002854.3 19 - 20 2512 97
GenBank mRNA AA523002.1 - 1 - 2 97
AF046785.1 - 19 - 20 2385 97
AF066858.1 - 19 - 20 2385 97
AK225904.1 - 19 - 20 2465 97
AK300580.1 - 18 - 19 2363 97
AK313206.1 - 19 - 20 2426 97
BC009895.2 - 19 - 20 2452 97
BC082229.1 - 19 - 20 2456 97
BC095850.1 - 19 - 20 2472 97
BC110791.1 - 19 - 20 2454 97
M14636.1 - 19 - 20 2498 97
M36807.1 - 8 - 9 942 97
Y15233.1 - 19 - 20 2436 97

View Full Product Details