The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002871.4 | NP_002862.2 | 1 - 2 | 289 | 64 |
GenBank mRNA | AK223538.1 | - | 1 - 2 | 289 | 64 |
AK311900.1 | - | 1 - 2 | 180 | 64 | |
BC018488.1 | - | 1 - 2 | 142 | 64 | |
BC037392.1 | - | 1 - 2 | 167 | 64 | |
BT007133.1 | - | 1 - 2 | 130 | 64 | |
S78873.1 | - | 1 - 2 | 142 | 64 | |
U74324.1 | - | 1 - 2 | 133 | 64 |