The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002894.2 | NP_002885.1 | 4 - 5 | 583 | 75 |
NM_203291.1 | NP_976036.1 | 4 - 5 | 553 | 75 | |
NM_203292.1 | NP_976037.1 | 4 - 5 | 634 | 75 | |
XM_005258325.2 | XP_005258382.1 | 4 - 5 | 514 | 75 | |
XM_005258326.3 | XP_005258383.1 | 3 - 4 | 350 | 75 | |
XM_006722519.2 | XP_006722582.1 | 6 - 7 | 655 | 75 | |
XM_006722520.2 | XP_006722583.1 | 5 - 6 | 591 | 75 | |
XM_006722521.2 | XP_006722584.1 | 4 - 5 | 651 | 75 | |
XM_011526132.2 | XP_011524434.1 | 5 - 6 | 602 | 75 | |
GenBank mRNA | AF043431.1 | - | 4 - 5 | 550 | 75 |
BC001170.1 | - | 4 - 5 | 739 | 75 | |
BC030590.1 | - | 4 - 5 | 381 | 75 | |
BU187672.1 | - | 4 - 5 | 523 | 75 | |
U72066.1 | - | 4 - 5 | 551 | 75 |