The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002945.3 | NP_002936.1 | 12 - 13 | 1359 | 91 |
GenBank mRNA | AB209732.1 | - | 12 - 13 | 1287 | 91 |
AK289704.1 | - | 12 - 13 | 1325 | 91 | |
BC018126.1 | - | 12 - 13 | 1287 | 91 | |
M63488.1 | - | 12 - 13 | 1313 | 91 |