The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001247997.1 | NP_001234926.1 | 3 - 4 | 814 | 65 |
NM_002956.2 | NP_002947.1 | 3 - 4 | 814 | 65 | |
NM_198240.1 | NP_937883.1 | 3 - 4 | 814 | 65 | |
XM_005253593.2 | XP_005253650.1 | 3 - 4 | 877 | 65 | |
XM_006719552.3 | XP_006719615.1 | 3 - 4 | 903 | 65 | |
XM_017019786.1 | XP_016875275.1 | 3 - 4 | 978 | 65 | |
XM_017019787.1 | XP_016875276.1 | 3 - 4 | 877 | 65 | |
XM_017019788.1 | XP_016875277.1 | 3 - 4 | 903 | 65 | |
XM_017019789.1 | XP_016875278.1 | 3 - 4 | 1062 | 65 | |
XM_017019790.1 | XP_016875279.1 | 3 - 4 | 804 | 65 | |
XM_017019791.1 | XP_016875280.1 | 3 - 4 | 903 | 65 | |
XM_017019792.1 | XP_016875281.1 | 3 - 4 | 903 | 65 | |
GenBank mRNA | BC047895.1 | - | 3 - 4 | 820 | 65 |
BC062543.1 | - | 2 - 3 | 770 | 65 | |
BC114213.1 | - | 2 - 3 | 729 | 65 | |
BC117209.1 | - | 2 - 3 | 701 | 65 | |
BC126305.1 | - | 2 - 3 | 701 | 65 | |
M97501.1 | - | 3 - 4 | 814 | 65 | |
X64838.1 | - | 3 - 4 | 791 | 65 |