Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CLEC11A
Entrez Gene ID
6320
Gene Name
C-type lectin domain family 11 member A
Gene Aliases
CLECSF3, LSLCL, P47, SCGF
Chromosome Location
Chr.19: 50723348 - 50725724 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_002975.2 NP_002966.1 2 - 3 514 102
GenBank mRNA AB009244.1 - 2 - 3 336 102
AF020044.1 - 2 - 3 515 102
AK314141.1 - 2 - 3 498 102
AY870345.1 - 2 - 3 339 102
BC005810.2 - 2 - 3 478 102
BT019673.1 - 2 - 3 336 102
D86586.1 - 2 - 3 475 102

View Full Product Details