The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001144757.1 | NP_001138229.1 | 2 - 3 | 347 | 78 |
NM_003020.3 | NP_003011.1 | 2 - 3 | 347 | 78 | |
GenBank mRNA | AK314965.1 | - | 2 - 3 | 340 | 78 |
BC005349.1 | - | 2 - 3 | 286 | 78 | |
BC093053.1 | - | 2 - 3 | 306 | 78 | |
BP377248.1 | - | 2 - 3 | 347 | 78 | |
CR541824.1 | - | 1 - 2 | 230 | 78 | |
Y00757.1 | - | 2 - 3 | 258 | 78 |