The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003052.4 | NP_003043.3 | 10 - 11 | 1277 | 75 |
XM_017009773.1 | XP_016865262.1 | 9 - 10 | 1220 | 75 | |
XM_017009774.1 | XP_016865263.1 | 3 - 4 | 375 | 75 | |
GenBank mRNA | AK096088.1 | - | 10 - 11 | 1256 | 75 |
BC035294.1 | - | 8 - 9 | 1464 | 75 | |
BC050385.1 | - | 10 - 11 | 1254 | 75 | |
BC053349.1 | - | 10 - 11 | 1264 | 75 | |
L13258.1 | - | 10 - 11 | 1254 | 75 |