Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC19A1
Entrez Gene ID
6573
Gene Name
solute carrier family 19 member 1
Gene Aliases
CHMD, FOLT, IFC1, REFC, RFC1
Chromosome Location
Chr.21: 45502517 - 45563143 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_011529698.2 XP_011528000.1 1 - 2 142 97

View Full Product Details