The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003098.2 | NP_003089.1 | 3 - 4 | 973 | 107 |
XM_005260517.1 | XP_005260574.1 | 3 - 4 | 934 | 107 | |
XM_011529007.1 | XP_011527309.1 | 3 - 4 | 934 | 107 | |
XM_011529008.1 | XP_011527310.1 | 3 - 4 | 934 | 107 | |
GenBank mRNA | AK095942.1 | - | 3 - 4 | 533 | 107 |
AK291994.1 | - | 3 - 4 | 844 | 107 | |
AK301800.1 | - | 3 - 4 | 704 | 107 | |
BC026215.2 | - | 3 - 4 | 813 | 107 | |
BC113813.1 | - | 1 - 2 | 121 | 107 | |
S81737.1 | - | 3 - 4 | 833 | 107 | |
U40571.1 | - | 3 - 4 | 738 | 107 |