The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001326542.1 | NP_001313471.1 | 4 - 5 | 2119 | 110 |
NM_001326543.1 | NP_001313472.1 | 4 - 5 | 2090 | 110 | |
NM_003112.4 | NP_003103.2 | 4 - 5 | 2124 | 110 | |
XM_005249828.2 | XP_005249885.1 | 4 - 5 | 2078 | 110 | |
XM_005249829.3 | XP_005249886.1 | 4 - 5 | 2058 | 110 | |
XM_011515486.2 | XP_011513788.1 | 4 - 5 | 2058 | 110 | |
GenBank mRNA | AK289728.1 | - | 4 - 5 | 2124 | 110 |
BC109300.1 | - | 4 - 5 | 2030 | 110 | |
BC109301.1 | - | 4 - 5 | 1996 | 110 | |
X68561.1 | - | 4 - 5 | 2087 | 110 |