The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003186.3 | NP_003177.2 | 1 - 2 | 113 | 82 |
GenBank mRNA | AK223002.1 | - | 1 - 2 | 67 | 82 |
AK314043.1 | - | 1 - 2 | 62 | 82 | |
BC010946.1 | - | 1 - 2 | 66 | 82 | |
BI830888.1 | - | 1 - 2 | 121 | 82 | |
M95787.1 | - | 1 - 2 | 67 | 82 |