The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003262.3 | NP_003253.1 | 1 - 2 | 68 | 98 |
GenBank mRNA | BC003677.1 | - | 1 - 2 | 60 | 98 |
BC007040.1 | - | 1 - 2 | 60 | 98 | |
BC012035.1 | - | 1 - 2 | 52 | 98 | |
BX475495.1 | - | 1 - 2 | 69 | 98 | |
BX648539.1 | - | 1 - 2 | 52 | 98 | |
D87127.1 | - | 1 - 2 | 49 | 98 | |
KC005990.1 | - | 1 - 2 | 37 | 98 | |
U93239.1 | - | 1 - 2 | 49 | 98 |