The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003342.4 | NP_003333.1 | 1 - 2 | 406 | 62 |
GenBank mRNA | AK313059.1 | - | 1 - 2 | 253 | 62 |
BC002775.2 | - | 1 - 2 | 305 | 62 | |
BC026288.1 | - | 1 - 2 | 214 | 62 | |
BG703127.1 | - | 1 - 2 | 378 | 62 | |
BI601205.1 | - | 1 - 2 | 417 | 62 | |
BT007416.1 | - | 1 - 2 | 48 | 62 | |
BU161770.1 | - | 1 - 2 | 347 | 62 | |
CD250951.1 | - | 1 - 2 | 383 | 62 | |
D78514.1 | - | 1 - 2 | 66 | 62 |