The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000097.5 | NP_000088.3 | 5 - 6 | 1277 | 118 |
GenBank mRNA | AK223481.1 | - | 5 - 6 | 1207 | 118 |
AK290140.1 | - | 5 - 6 | 1277 | 118 | |
BC017210.2 | - | 5 - 6 | 1228 | 118 | |
BC023551.2 | - | 5 - 6 | 1263 | 118 | |
BC023554.2 | - | 5 - 6 | 1224 | 118 | |
D16611.1 | - | 5 - 6 | 963 | 118 | |
Z28409.1 | - | 5 - 6 | 870 | 118 |