The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000104.3 | NP_000095.2 | 2 - 3 | 1442 | 118 |
GenBank mRNA | AK297039.1 | - | 2 - 3 | 1025 | 118 |
AK312686.1 | - | 2 - 3 | 1444 | 118 | |
BC012049.1 | - | 2 - 3 | 1395 | 118 | |
BT019979.1 | - | 1 - 2 | 1039 | 118 | |
BT020001.1 | - | 1 - 2 | 1039 | 118 | |
U03688.1 | - | 2 - 3 | 1385 | 118 |