The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000180.3 | NP_000171.1 | 19 - 20 | 3410 | 135 |
GenBank mRNA | BM931704.1 | - | 2 - 3 | 158 | 135 |
M92432.1 | - | 19 - 20 | 3409 | 135 |