Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 28399775-28400283 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SARM1
Entrez Gene ID
23098
Gene Name
sterile alpha and TIR motif containing 1
Gene Aliases
MyD88-5, SAMD2, SARM
UniGene
Hs.743510
Gene Chromosome Location
Chr.17: 28371662-28401045 on Build GRCh38
Transcripts
NM_015077.3 NM_001242366.2 NM_080669.5 XM_005277786.3 XM_017024110.1

Gene Symbol
SLC46A1
Entrez Gene ID
113235
Gene Name
solute carrier family 46 member 1
Gene Aliases
G21, HCP1, PCFT
UniGene
Hs.446689
Gene Chromosome Location
Chr.17: 28394642-28406630 on Build GRCh38
Transcripts
NM_015077.3 NM_001242366.2 NM_080669.5 XM_005277786.3 XM_017024110.1

SNP Detail

SNP ID Chromosome Location
rs1026576413 Chr.17: 28399850-28399850 on Build GRCh38
rs895263586 Chr.17: 28400177-28400177 on Build GRCh38
rs954685664 Chr.17: 28400249-28400249 on Build GRCh38
rs910327497 Chr.17: 28400011-28400011 on Build GRCh38
rs987647468 Chr.17: 28399938-28399938 on Build GRCh38
rs1005551911 Chr.17: 28400186-28400186 on Build GRCh38
rs963327687 Chr.17: 28399809-28399809 on Build GRCh38
rs531937212 Chr.17: 28399886-28399886 on Build GRCh38
rs1038380293 Chr.17: 28400191-28400191 on Build GRCh38
rs199646933 Chr.17: 28400162-28400162 on Build GRCh38
rs945639396 Chr.17: 28400118-28400118 on Build GRCh38
rs770950918 Chr.17: 28400046-28400046 on Build GRCh38
rs41297119 Chr.17: 28399917-28399917 on Build GRCh38
rs782455755 Chr.17: 28400037-28400037 on Build GRCh38
rs1042531127 Chr.17: 28400122-28400122 on Build GRCh38
rs993303948 Chr.17: 28399811-28399811 on Build GRCh38
rs774508059 Chr.17: 28400123-28400123 on Build GRCh38
rs1026126268 Chr.17: 28400236-28400236 on Build GRCh38
rs915492044 Chr.17: 28400111-28400111 on Build GRCh38
rs989140679 Chr.17: 28400110-28400110 on Build GRCh38
rs41297117 Chr.17: 28400155-28400155 on Build GRCh38
rs71372377 Chr.17: 28399843-28399843 on Build GRCh38
rs993836632 Chr.17: 28400235-28400235 on Build GRCh38
rs543888065 Chr.17: 28399934-28399934 on Build GRCh38
rs372715380 Chr.17: 28400044-28400044 on Build GRCh38
rs190910424 Chr.17: 28400169-28400169 on Build GRCh38
rs564435609 Chr.17: 28399862-28399862 on Build GRCh38
rs41297115 Chr.17: 28400220-28400220 on Build GRCh38

View Full Product Details