Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC12A1
Entrez Gene ID
6557
Gene Name
solute carrier family 12 member 1
Gene Aliases
BSC1, NKCC2
Chromosome Location
Chr.15: 48206301 - 48304078 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000338.2 NP_000329.2 13 - 14 1904 65
NM_001184832.1 NP_001171761.1 13 - 14 1904 65
XM_005254606.1 XP_005254663.1 13 - 14 1904 65
GenBank mRNA AK295375.1 - 12 - 13 1357 65
AK298275.1 - 8 - 9 1123 65
AK298312.1 - 13 - 14 1903 65
BX647067.1 - 8 - 9 5827 65
BX647484.1 - 9 - 10 5811 65
EF559316.2 - 12 - 13 1702 65
U58130.1 - 12 - 13 1707 65

View Full Product Details