The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000341.3 | NP_000332.2 | 6 - 7 | 1216 | 92 |
XM_011533047.2 | XP_011531349.1 | 6 - 7 | 1216 | 92 | |
GenBank mRNA | AB033549.1 | - | 6 - 7 | 1197 | 92 |
AK091932.1 | - | 7 - 8 | 1082 | 92 | |
AK092000.1 | - | 3 - 4 | 1322 | 92 | |
AK223146.1 | - | 6 - 7 | 1178 | 92 | |
AK289636.1 | - | 6 - 7 | 1194 | 92 | |
AK309145.1 | - | 6 - 7 | 1210 | 92 | |
AK313157.1 | - | 6 - 7 | 1210 | 92 | |
BC022386.1 | - | 6 - 7 | 1214 | 92 | |
BC093624.1 | - | 6 - 7 | 1175 | 92 | |
BC093626.1 | - | 6 - 7 | 1170 | 92 | |
DQ023512.1 | - | 3 - 4 | 1322 | 92 | |
DQ023515.1 | - | 6 - 7 | 1138 | 92 | |
DQ023516.1 | - | 6 - 7 | 1138 | 92 | |
DQ023517.1 | - | 6 - 7 | 1138 | 92 | |
KU178335.1 | - | 6 - 7 | 1138 | 92 | |
L11696.1 | - | 6 - 7 | 1181 | 92 | |
M95548.1 | - | 6 - 7 | 1182 | 92 |