The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000403.3 | NP_000394.2 | 6 - 7 | 760 | 154 |
NM_001008216.1 | NP_001008217.1 | 6 - 7 | 695 | 154 | |
NM_001127621.1 | NP_001121093.1 | 5 - 6 | 736 | 154 | |
GenBank mRNA | AK057302.1 | - | 4 - 5 | 931 | 154 |
AK314397.1 | - | 5 - 6 | 708 | 154 | |
AL530475.3 | - | 6 - 7 | 694 | 154 | |
BC001273.1 | - | 5 - 6 | 616 | 154 | |
BC050685.2 | - | 6 - 7 | 619 | 154 | |
DQ233668.1 | - | 4 - 5 | 526 | 154 | |
L41668.1 | - | 6 - 7 | 619 | 154 |