The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000418.3 | NP_000409.1 | 4 - 5 | 473 | 70 |
NM_001257406.1 | NP_001244335.1 | 3 - 4 | 340 | 70 | |
NM_001257407.1 | NP_001244336.1 | 4 - 5 | 584 | 70 | |
NM_001257997.1 | NP_001244926.1 | 3 - 4 | 340 | 70 | |
XM_011545825.1 | XP_011544127.1 | 3 - 4 | 378 | 70 | |
XM_011545826.2 | XP_011544128.1 | 4 - 5 | 429 | 70 | |
XM_011545827.2 | XP_011544129.1 | 4 - 5 | 1668 | 70 | |
XM_011545829.1 | XP_011544131.1 | 4 - 5 | 459 | 70 | |
XM_011545830.2 | XP_011544132.1 | 4 - 5 | 1669 | 70 | |
XM_011545831.1 | XP_011544133.1 | 3 - 4 | 378 | 70 | |
XM_011545832.1 | XP_011544134.1 | 3 - 4 | 311 | 70 | |
XM_011545833.2 | XP_011544135.1 | 2 - 3 | 229 | 70 | |
XM_017023211.1 | XP_016878700.1 | 3 - 4 | 378 | 70 | |
GenBank mRNA | AK223591.1 | - | 4 - 5 | 391 | 70 |
AK303255.1 | - | 4 - 5 | 559 | 70 | |
AK314863.1 | - | 3 - 4 | 325 | 70 | |
BC136365.1 | - | 3 - 4 | 372 | 70 | |
BC151131.1 | - | 3 - 4 | 372 | 70 | |
DA492909.1 | - | 3 - 4 | 341 | 70 | |
X52425.1 | - | 4 - 5 | 385 | 70 |