The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000452.2 | NP_000443.1 | 4 - 5 | 1358 | 98 |
GenBank mRNA | BC130521.1 | - | 4 - 5 | 794 | 98 |
BC130523.1 | - | 4 - 5 | 794 | 98 | |
U10417.1 | - | 4 - 5 | 1359 | 98 |