The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000495.4 | NP_000486.1 | 1 - 2 | 363 | 78 |
NM_033380.2 | NP_203699.1 | 1 - 2 | 363 | 78 | |
GenBank mRNA | BC151846.1 | - | 1 - 2 | 325 | 78 |
KM282618.1 | - | 1 - 2 | 363 | 78 | |
M90464.1 | - | 1 - 2 | 283 | 78 |