Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 58734112-58734554 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RAD51C
Entrez Gene ID
5889
Gene Name
RAD51 paralog C
Gene Aliases
BROVCA3, FANCO, R51H3, RAD51L2
UniGene
Hs.412587
Gene Chromosome Location
Chr.17: 58692538-58734342 on Build GRCh38
Transcripts
NM_058216.2 NR_103872.1 XM_006722001.3 XM_006722002.3 XM_006722004.3 XM_006722005.3 XM_011525092.2 XM_011525093.2 XM_011525094.2 XM_017024914.1 XM_017024915.1 XM_017024916.1 XM_017024917.1 XM_017024918.1 XM_017024919.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
981988 c.1070T>C p.L357S Chr.17: 58734161-58734161 on Build GRCh38
346482 c.1068A>T p.S356S Chr.17: 58734159-58734159 on Build GRCh38
1384847 c.1045A>G p.T349A Chr.17: 58734136-58734136 on Build GRCh38
981989 c.1103G>A p.R368Q Chr.17: 58734194-58734194 on Build GRCh38
6921743 c.1067C>G p.S356* Chr.17: 58734158-58734158 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs954599032 Chr.17: 58734454-58734454 on Build GRCh38
rs763504056 Chr.17: 58734271-58734271 on Build GRCh38
rs756744016 Chr.17: 58734199-58734199 on Build GRCh38
rs896048860 Chr.17: 58734299-58734299 on Build GRCh38
rs776541633 Chr.17: 58734307-58734307 on Build GRCh38
rs2611784 Chr.17: 58734390-58734390 on Build GRCh38
rs1029636375 Chr.17: 58734452-58734452 on Build GRCh38
rs185890675 Chr.17: 58734436-58734436 on Build GRCh38
rs772954427 Chr.17: 58734519-58734519 on Build GRCh38
rs768965452 Chr.17: 58734244-58734244 on Build GRCh38
rs1056821626 Chr.17: 58734279-58734279 on Build GRCh38
rs577852020 Chr.17: 58734188-58734188 on Build GRCh38
rs925843783 Chr.17: 58734420-58734420 on Build GRCh38
rs772459278 Chr.17: 58734259-58734259 on Build GRCh38
rs761770500 Chr.17: 58734152-58734152 on Build GRCh38
rs748051956 Chr.17: 58734251-58734251 on Build GRCh38
rs864622632 Chr.17: 58734217-58734217 on Build GRCh38
rs559647198 Chr.17: 58734180-58734180 on Build GRCh38
rs374605909 Chr.17: 58734253-58734253 on Build GRCh38
rs181365564 Chr.17: 58734262-58734262 on Build GRCh38
rs545024029 Chr.17: 58734219-58734219 on Build GRCh38
rs781286948 Chr.17: 58734232-58734232 on Build GRCh38
rs587782426 Chr.17: 58734154-58734154 on Build GRCh38
rs888598147 Chr.17: 58734354-58734354 on Build GRCh38
rs373170458 Chr.17: 58734200-58734200 on Build GRCh38
rs587782565 Chr.17: 58734181-58734181 on Build GRCh38
rs910482318 Chr.17: 58734351-58734351 on Build GRCh38
rs587782449 Chr.17: 58734187-58734187 on Build GRCh38
rs377262060 Chr.17: 58734220-58734220 on Build GRCh38
rs934477487 Chr.17: 58734498-58734498 on Build GRCh38
rs587780253 Chr.17: 58734193-58734193 on Build GRCh38
rs548687636 Chr.17: 58734317-58734317 on Build GRCh38
rs747899252 Chr.17: 58734383-58734383 on Build GRCh38
rs1047375446 Chr.17: 58734316-58734316 on Build GRCh38
rs865988490 Chr.17: 58734159-58734159 on Build GRCh38
rs2611785 Chr.17: 58734403-58734403 on Build GRCh38
rs755838887 Chr.17: 58734185-58734185 on Build GRCh38
rs191215184 Chr.17: 58734479-58734479 on Build GRCh38
rs201000407 Chr.17: 58734153-58734153 on Build GRCh38
rs766809303 Chr.17: 58734178-58734178 on Build GRCh38
rs773843495 Chr.17: 58734158-58734158 on Build GRCh38
rs876659052 Chr.17: 58734226-58734226 on Build GRCh38
rs562349220 Chr.17: 58734348-58734348 on Build GRCh38
rs998155191 Chr.17: 58734422-58734422 on Build GRCh38
rs786203771 Chr.17: 58734146-58734146 on Build GRCh38
rs776823687 Chr.17: 58734137-58734137 on Build GRCh38
rs1006171166 Chr.17: 58734370-58734370 on Build GRCh38
rs786202691 Chr.17: 58734210-58734210 on Build GRCh38
rs943261580 Chr.17: 58734358-58734358 on Build GRCh38
rs28363336 Chr.17: 58734247-58734247 on Build GRCh38
rs763424319 Chr.17: 58734168-58734168 on Build GRCh38
rs863224805 Chr.17: 58734183-58734183 on Build GRCh38
rs899920856 Chr.17: 58734397-58734397 on Build GRCh38
rs769937101 Chr.17: 58734258-58734258 on Build GRCh38
rs117954900 Chr.17: 58734505-58734505 on Build GRCh38
rs148537339 Chr.17: 58734140-58734140 on Build GRCh38
rs758223998 Chr.17: 58734209-58734209 on Build GRCh38
rs533020255 Chr.17: 58734512-58734512 on Build GRCh38
rs560599686 Chr.17: 58734323-58734323 on Build GRCh38
rs895514056 Chr.17: 58734167-58734167 on Build GRCh38

View Full Product Details