The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000712.3 | NP_000703.2 | 3 - 4 | 216 | 128 |
NM_001253823.1 | NP_001240752.1 | 4 - 5 | 310 | 128 | |
XM_011515474.2 | XP_011513776.1 | 3 - 4 | 218 | 128 | |
XM_017012520.1 | XP_016868009.1 | 3 - 4 | 225 | 128 | |
GenBank mRNA | AK291862.1 | - | 4 - 5 | 297 | 128 |
BC005902.1 | - | 3 - 4 | 209 | 128 | |
BC008456.1 | - | 3 - 4 | 209 | 128 | |
BM710258.1 | - | 3 - 4 | 216 | 128 | |
U34877.1 | - | 3 - 4 | 189 | 128 | |
X93086.1 | - | 3 - 4 | 193 | 128 |