Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 46031957-46032464 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KANSL1
Entrez Gene ID
284058
Gene Name
KAT8 regulatory NSL complex subunit 1
Gene Aliases
CENP-36, KDVS, KIAA1267, MSL1v1, NSL1, hMSL1v1
UniGene
Hs.648744
Gene Chromosome Location
Chr.17: 46029916-46225374 on Build GRCh38
Transcripts
NM_001193465.1 NM_001193466.1 NM_015443.3 XM_006721823.1 XM_006721824.3 XM_011524628.2 XM_011524629.2 XM_011524630.2 XM_011524631.2 XM_011524632.2 XM_017024488.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
980236 c.3052A>G p.T1018A Chr.17: 46032085-46032085 on Build GRCh38
4485182 c.2901C>T p.P967P Chr.17: 46032236-46032236 on Build GRCh38
4485604 c.2951C>T p.S984F Chr.17: 46032186-46032186 on Build GRCh38
4485611 c.2952C>T p.S984S Chr.17: 46032185-46032185 on Build GRCh38
5989199 c.2896A>C p.T966P Chr.17: 46032241-46032241 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1057469020 Chr.17: 46032191-46032191 on Build GRCh38
rs748843892 Chr.17: 46032107-46032107 on Build GRCh38
rs768133827 Chr.17: 46032332-46032332 on Build GRCh38
rs190142867 Chr.17: 46032308-46032308 on Build GRCh38
rs755795005 Chr.17: 46032185-46032185 on Build GRCh38
rs752833970 Chr.17: 46032064-46032064 on Build GRCh38
rs370056222 Chr.17: 46032389-46032389 on Build GRCh38
rs761823833 Chr.17: 46032350-46032350 on Build GRCh38
rs781056926 Chr.17: 46032081-46032081 on Build GRCh38
rs758333951 Chr.17: 46032067-46032067 on Build GRCh38
rs770177501 Chr.17: 46032237-46032237 on Build GRCh38
rs904483649 Chr.17: 46032334-46032334 on Build GRCh38
rs946207471 Chr.17: 46031983-46031983 on Build GRCh38
rs753548997 Chr.17: 46032203-46032203 on Build GRCh38
rs763570104 Chr.17: 46032251-46032251 on Build GRCh38
rs772500977 Chr.17: 46032012-46032012 on Build GRCh38
rs768555471 Chr.17: 46032241-46032241 on Build GRCh38
rs780672258 Chr.17: 46031998-46031998 on Build GRCh38
rs1010687863 Chr.17: 46032263-46032263 on Build GRCh38
rs7220988 Chr.17: 46032108-46032108 on Build GRCh38
rs1076222 Chr.17: 46032403-46032403 on Build GRCh38
rs1037585739 Chr.17: 46032135-46032135 on Build GRCh38
rs751551522 Chr.17: 46032072-46032072 on Build GRCh38
rs974482953 Chr.17: 46032035-46032035 on Build GRCh38
rs376138960 Chr.17: 46032017-46032017 on Build GRCh38
rs774866655 Chr.17: 46032321-46032321 on Build GRCh38
rs148203818 Chr.17: 46032117-46032117 on Build GRCh38
rs747564871 Chr.17: 46032110-46032110 on Build GRCh38
rs781514778 Chr.17: 46032224-46032224 on Build GRCh38
rs747465838 Chr.17: 46032212-46032212 on Build GRCh38
rs766288923 Chr.17: 46032329-46032329 on Build GRCh38
rs367992334 Chr.17: 46032351-46032351 on Build GRCh38
rs778490119 Chr.17: 46032211-46032211 on Build GRCh38
rs146848167 Chr.17: 46032188-46032188 on Build GRCh38
rs905710793 Chr.17: 46032180-46032180 on Build GRCh38
rs143018360 Chr.17: 46032109-46032109 on Build GRCh38
rs777045712 Chr.17: 46032112-46032112 on Build GRCh38
rs376488097 Chr.17: 46031997-46031997 on Build GRCh38
rs764260806 Chr.17: 46032146-46032146 on Build GRCh38
rs140588825 Chr.17: 46032425-46032425 on Build GRCh38
rs767403167 Chr.17: 46032179-46032179 on Build GRCh38
rs201117362 Chr.17: 46032038-46032038 on Build GRCh38
rs769035708 Chr.17: 46032253-46032253 on Build GRCh38
rs763978952 Chr.17: 46032071-46032071 on Build GRCh38
rs536638673 Chr.17: 46032377-46032377 on Build GRCh38
rs376635152 Chr.17: 46032276-46032276 on Build GRCh38
rs976874519 Chr.17: 46032405-46032405 on Build GRCh38
rs754725774 Chr.17: 46032339-46032339 on Build GRCh38
rs767599124 Chr.17: 46032272-46032272 on Build GRCh38
rs774560239 Chr.17: 46032252-46032252 on Build GRCh38
rs1041841138 Chr.17: 46032411-46032411 on Build GRCh38
rs745540054 Chr.17: 46031999-46031999 on Build GRCh38
rs778272918 Chr.17: 46032349-46032349 on Build GRCh38
rs746273618 Chr.17: 46032227-46032227 on Build GRCh38
rs371484505 Chr.17: 46032152-46032152 on Build GRCh38
rs750288937 Chr.17: 46032183-46032183 on Build GRCh38
rs748649772 Chr.17: 46032008-46032008 on Build GRCh38
rs763252158 Chr.17: 46032240-46032240 on Build GRCh38
rs765297359 Chr.17: 46032055-46032055 on Build GRCh38
rs779686052 Chr.17: 46032106-46032106 on Build GRCh38
rs771527693 Chr.17: 46032218-46032218 on Build GRCh38
rs769373626 Chr.17: 46032000-46032000 on Build GRCh38
rs145863194 Chr.17: 46032084-46032084 on Build GRCh38
rs776773399 Chr.17: 46032026-46032026 on Build GRCh38
rs895889126 Chr.17: 46032229-46032229 on Build GRCh38
rs201824489 Chr.17: 46032004-46032004 on Build GRCh38
rs775516828 Chr.17: 46032129-46032129 on Build GRCh38
rs753364053 Chr.17: 46032333-46032333 on Build GRCh38
rs773591156 Chr.17: 46032015-46032015 on Build GRCh38
rs754387068 Chr.17: 46032210-46032210 on Build GRCh38
rs138490347 Chr.17: 46032099-46032099 on Build GRCh38
rs573432386 Chr.17: 46032126-46032126 on Build GRCh38
rs993250448 Chr.17: 46032322-46032322 on Build GRCh38
rs761819132 Chr.17: 46032153-46032153 on Build GRCh38
rs762905143 Chr.17: 46032134-46032134 on Build GRCh38
rs147378906 Chr.17: 46032265-46032265 on Build GRCh38
rs796052588 Chr.17: 46032186-46032186 on Build GRCh38
rs775643555 Chr.17: 46032239-46032239 on Build GRCh38
rs756958045 Chr.17: 46032075-46032075 on Build GRCh38

View Full Product Details