Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CYP4A11
Entrez Gene ID
1579
Gene Name
cytochrome P450 family 4 subfamily A member 11
Gene Aliases
CP4Y, CYP4A2, CYP4AII
Chromosome Location
Chr.1: 46929174 - 46941484 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000778.3 NP_000769.2 11 - 12 1415 115
NM_001319155.1 NP_001306084.1 11 - 12 1319 115
NR_134988.1 9 - 10 1069 115
NR_134989.1 10 - 11 1260 115
NR_134990.1 10 - 11 1254 115
NR_134991.1 10 - 11 1241 115
NR_134992.1 7 - 8 870 115
NR_134993.1 8 - 9 1004 115
NR_134994.1 10 - 11 1276 115
XM_017000464.1 XP_016855953.1 9 - 10 1113 115
XM_017000465.1 XP_016855954.1 11 - 12 1334 115
GenBank mRNA CD013974.1 - 11 - 12 115
CD013975.1 - 9 - 10 115
CD013976.1 - 10 - 11 115
CD013977.1 - 10 - 11 115
CD013978.1 - 10 - 11 115
CD013979.1 - 7 - 8 115
CD013980.1 - 8 - 9 115
CD014132.1 - 10 - 11 115
D13705.1 - 11 - 12 118
D26481.1 - 11 - 12 1379 115
L04751.1 - 11 - 12 115
S67580.1 - 11 - 12 1405 115
S67581.1 - 11 - 12 1405 115

Gene Symbol
CYP4A22
Entrez Gene ID
284541
Gene Name
cytochrome P450 family 4 subfamily A member 22
Gene Aliases
-
Chromosome Location
Chr.1: 47137425 - 47149738 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001010969.3 NP_001010969.2 11 - 12 1425 115
XM_005270767.3 XP_005270824.1 9 - 10 1113 115
GenBank mRNA AY280371.1 - 11 - 12 1415 115
AY280372.1 - 11 - 12 1364 115
BC148248.1 - 11 - 12 1425 115

View Full Product Details