Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DRD3
Entrez Gene ID
1814
Gene Name
dopamine receptor D3
Gene Aliases
D3DR, ETM1, FET1
Chromosome Location
Chr.3: 114127623 - 114199407 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000796.5 NP_000787.2 5 - 6 1155 84
NM_001282563.2 NP_001269492.1 6 - 7 1014 84
NM_001290809.1 NP_001277738.1 6 - 7 1147 84
NM_033663.5 NP_387512.3 5 - 6 1155 84
GenBank mRNA AK292309.1 - 6 - 7 1014 84
BC095510.1 - 4 - 5 725 84
BC128122.1 - 6 - 7 1069 84
BC128123.1 - 5 - 6 801 84
KJ534829.1 - 4 - 5 724 84
KJ535032.1 - 3 - 4 611 84
L20469.1 - 4 - 5 724 84
U32499.1 - 4 - 5 724 84

View Full Product Details