The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000834.3 | NP_000825.2 | 4 - 5 | 1306 | 78 |
XM_011520628.2 | XP_011518930.1 | 5 - 6 | 2099 | 78 | |
XM_011520629.2 | XP_011518931.1 | 6 - 7 | 1906 | 78 | |
XM_017019219.1 | XP_016874708.1 | 5 - 6 | 1654 | 78 | |
GenBank mRNA | AB208850.1 | - | 1 - 2 | 85 | 78 |
BC113618.1 | - | 4 - 5 | 1205 | 78 | |
BC113620.1 | - | 4 - 5 | 1205 | 78 | |
U11287.1 | - | 4 - 5 | 1337 | 78 | |
U88963.1 | - | 4 - 5 | 1306 | 78 | |
U90278.1 | - | 4 - 5 | 1336 | 78 |