Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GRM4
Entrez Gene ID
2914
Gene Name
glutamate metabotropic receptor 4
Gene Aliases
GPRC1D, MGLUR4, mGlu4
Chromosome Location
Chr.6: 34018643 - 34155622 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000841.3 NP_000832.1 4 - 5 1377 96
NM_001256809.2 NP_001243738.1 3 - 4 901 96
NM_001256811.2 NP_001243740.1 4 - 5 1377 96
NM_001256812.2 NP_001243741.1 3 - 4 461 96
NM_001256813.2 NP_001243742.1 4 - 5 685 96
NM_001282847.1 NP_001269776.1 3 - 4 1160 96
XM_017010790.1 XP_016866279.1 4 - 5 1315 96
XM_017010791.1 XP_016866280.1 4 - 5 1370 96
XM_017010792.1 XP_016866281.1 3 - 4 914 96
XM_017010793.1 XP_016866282.1 3 - 4 665 96
GenBank mRNA AK122836.1 - 1 - 2 1351 96
AK122982.1 - 4 - 5 670 96
AK126746.1 - 2 - 3 430 96
AK131536.1 - 3 - 4 901 96
AK289524.1 - - 1695 96
AK289534.1 - 4 - 5 1320 96
AK289563.1 - 2 - 3 234 96
AK293913.1 - 3 - 4 461 96
AK293949.1 - 3 - 4 487 96
AK312300.1 - 3 - 4 1115 96
BC130526.1 - 3 - 4 1035 96
BC130528.1 - 3 - 4 1035 96
BC144057.1 - 3 - 4 1035 96
EU432124.1 - 3 - 4 876 96
U92457.1 - 3 - 4 901 96
X80818.1 - 3 - 4 1046 96

View Full Product Details