The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000842.4 | NP_000833.1 | 7 - 8 | 2084 | 80 |
NM_001143831.2 | NP_001137303.1 | 6 - 7 | 1886 | 80 | |
XM_006718828.3 | XP_006718891.1 | 7 - 8 | 2082 | 80 | |
XM_011542792.1 | XP_011541094.1 | 7 - 8 | 2084 | 80 | |
XM_017017627.1 | XP_016873116.1 | 7 - 8 | 2082 | 80 | |
GenBank mRNA | AK291362.1 | - | 7 - 8 | 2054 | 80 |
AY608336.1 | - | 6 - 7 | 1686 | 80 | |
D28538.1 | - | 6 - 7 | 1836 | 80 | |
D28539.1 | - | 6 - 7 | 1836 | 80 | |
FJ588505.1 | - | 6 - 7 | 1686 | 80 | |
FJ588506.1 | - | 6 - 7 | 1686 | 80 | |
FJ588507.1 | - | 6 - 7 | 1686 | 80 | |
FJ668670.1 | - | 6 - 7 | 1686 | 80 |