The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000680.3 | NP_000671.2 | 2 - 3 | 1882 | 112 |
NM_033302.3 | NP_150645.2 | 1 - 2 | 1574 | 112 | |
NM_033303.4 | NP_150646.3 | 1 - 2 | 1574 | 112 | |
NM_033304.3 | NP_150647.2 | 1 - 2 | 1574 | 112 | |
XM_006716292.3 | XP_006716355.1 | 2 - 3 | 1909 | 112 | |
XM_006716293.3 | XP_006716356.1 | 2 - 3 | 1909 | 112 | |
XM_017013094.1 | XP_016868583.1 | 3 - 4 | 5469 | 112 | |
XM_017013095.1 | XP_016868584.1 | 3 - 4 | 5492 | 112 | |
GenBank mRNA | AF013261.1 | - | 1 - 2 | 1088 | 112 |
AF395806.1 | - | 1 - 2 | 893 | 112 | |
AK289548.1 | - | 2 - 3 | 1864 | 112 | |
BC095512.1 | - | 1 - 2 | 900 | 112 | |
D25235.1 | - | 1 - 2 | 1324 | 112 | |
D32201.1 | - | 1 - 2 | 1324 | 112 | |
D32202.1 | - | 1 - 2 | 1324 | 112 | |
L31774.1 | - | 1 - 2 | 888 | 112 | |
S76001.1 | - | 1 - 2 | 287 | 112 | |
U02569.1 | - | 1 - 2 | 1312 | 112 |