The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001290223.1 | NP_001277152.1 | 24 - 25 | 2627 | 71 |
NM_001380.4 | NP_001371.1 | 24 - 25 | 2564 | 71 | |
XM_011539422.2 | XP_011537724.1 | 24 - 25 | 2605 | 71 | |
XM_011539424.1 | XP_011537726.1 | 24 - 25 | 2515 | 71 | |
XM_017015813.1 | XP_016871302.1 | 24 - 25 | 2537 | 71 | |
XM_017015814.1 | XP_016871303.1 | 24 - 25 | 2599 | 71 | |
XM_017015815.1 | XP_016871304.1 | 24 - 25 | 2622 | 71 | |
XM_017015816.1 | XP_016871305.1 | 7 - 8 | 785 | 71 | |
XM_017015817.1 | XP_016871306.1 | 24 - 25 | 2599 | 71 | |
GenBank mRNA | BC146857.1 | - | 24 - 25 | 2517 | 71 |
D50857.1 | - | 24 - 25 | 2478 | 71 |