Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PHLDA2
Entrez Gene ID
7262
Gene Name
pleckstrin homology like domain family A member 2
Gene Aliases
BRW1C, BWR1C, HLDA2, IPL, TSSC3
Chromosome Location
Chr.11: 2928273 - 2929420 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_003311.3 NP_003302.1 1 - 2 527 121
GenBank mRNA AF001294.1 - 1 - 2 527 121
AF019953.1 - 1 - 2 488 121
AF035444.1 - 1 - 2 527 121
AK223027.1 - 1 - 2 527 121
BC005034.1 - 1 - 2 517 121

View Full Product Details