The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001286974.1 | NP_001273903.1 | 11 - 12 | 1745 | 85 |
NM_003798.3 | NP_003789.1 | 11 - 12 | 1745 | 85 | |
XM_005252291.3 | XP_005252348.1 | 11 - 12 | 1713 | 85 | |
XM_017015250.1 | XP_016870739.1 | 10 - 11 | 1465 | 85 | |
XM_017015251.1 | XP_016870740.1 | 10 - 11 | 1465 | 85 | |
GenBank mRNA | AF006070.1 | - | 11 - 12 | 1635 | 85 |
AF030233.1 | - | 11 - 12 | 1637 | 85 | |
AF080071.1 | - | 11 - 12 | 1632 | 85 | |
AF135021.1 | - | 11 - 12 | 1636 | 85 | |
AK022539.1 | - | 10 - 11 | 1427 | 85 | |
AK022834.1 | - | 11 - 12 | 1678 | 85 | |
AK022926.1 | - | 11 - 12 | 1650 | 85 | |
AK223230.1 | - | 11 - 12 | 1650 | 85 | |
AK314677.1 | - | 11 - 12 | 1649 | 85 | |
AL050016.1 | - | 4 - 5 | 457 | 85 | |
BC117208.1 | - | 11 - 12 | 1614 | 85 | |
BC117234.1 | - | 11 - 12 | 1614 | 85 | |
U97067.1 | - | 11 - 12 | 1636 | 85 |