The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001143830.1 | NP_001137302.1 | 5 - 6 | 779 | 123 |
NM_005256.3 | NP_005247.1 | 4 - 5 | 570 | 123 | |
NM_177553.2 | NP_808221.1 | 5 - 6 | 721 | 123 | |
XM_011519971.2 | XP_011518273.1 | 5 - 6 | 779 | 123 | |
XM_011519972.2 | XP_011518274.1 | 5 - 6 | 2774 | 123 | |
XM_011519975.1 | XP_011518277.1 | 5 - 6 | 779 | 123 | |
XM_011519978.2 | XP_011518280.1 | 5 - 6 | 779 | 123 | |
XM_017017528.1 | XP_016873017.1 | 5 - 6 | 1471 | 123 | |
XM_017017529.1 | XP_016873018.1 | 5 - 6 | 616 | 123 | |
XM_017017530.1 | XP_016873019.1 | 5 - 6 | 809 | 123 | |
XM_017017531.1 | XP_016873020.1 | 5 - 6 | 630 | 123 | |
XM_017017532.1 | XP_016873021.1 | 5 - 6 | 779 | 123 | |
GenBank mRNA | AK313734.1 | - | 5 - 6 | 785 | 123 |
BC013326.1 | - | 6 - 7 | 791 | 123 | |
BC040470.1 | - | 5 - 6 | 721 | 123 | |
CR450285.1 | - | 4 - 5 | 473 | 123 | |
U95032.1 | - | 4 - 5 | 539 | 123 |