Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MAEA
Entrez Gene ID
10296
Gene Name
macrophage erythroblast attacher
Gene Aliases
EMLP, EMP, GID9, HLC-10, PIG5
Chromosome Location
Chr.4: 1289851 - 1340148 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001017405.2 NP_001017405.1 2 - 3 321 52
NM_001297431.1 NP_001284360.1 2 - 3 321 52
NM_001297432.1 NP_001284361.1 2 - 3 459 52
NM_001297433.1 NP_001284362.1 2 - 3 395 52
NM_005882.4 NP_005873.2 2 - 3 321 52
NR_123716.1 2 - 3 321 52
XM_006713849.3 XP_006713912.1 2 - 3 281 52
XM_006713850.2 XP_006713913.1 2 - 3 454 52
XM_011513376.1 XP_011511678.1 3 - 4 441 52
XM_017007636.1 XP_016863125.1 3 - 4 441 52
GenBank mRNA AF084928.1 - 3 - 4 269 52
AK001088.1 - 2 - 3 281 52
AK022515.1 - 2 - 3 290 52
AK022586.1 - 2 - 3 281 52
AK298944.1 - 2 - 3 375 52
AK300572.1 - 3 - 4 218 52
AK301155.1 - 2 - 3 289 52
AK308901.1 - 2 - 3 265 52
AY189687.1 - 2 - 3 234 52
AY236486.1 - 2 - 3 266 52
BC001225.1 - 2 - 3 288 52
BC006470.1 - 2 - 3 244 52
BT006957.1 - 2 - 3 225 52
DC299400.1 - 2 - 3 398 52
DC331820.1 - 2 - 3 322 52
DC400017.1 - 2 - 3 459 52

View Full Product Details