The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000215.3 | NP_000206.2 | 23 - 24 | 3310 | 95 |
XM_005259896.3 | XP_005259953.1 | 23 - 24 | 3349 | 95 | |
GenBank mRNA | U09607.1 | - | 22 - 23 | 3305 | 95 |