The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004590.3 | NP_004581.1 | 1 - 2 | 157 | 99 |
XM_005258020.4 | XP_005258077.1 | 3 - 4 | 439 | 99 | |
GenBank mRNA | AB007454.1 | - | 1 - 2 | 157 | 99 |
AF039955.1 | - | 1 - 2 | 148 | 99 | |
AF055467.1 | - | 1 - 2 | 115 | 99 | |
BC099662.3 | - | 1 - 2 | 122 | 99 | |
U91746.1 | - | 1 - 2 | 81 | 99 |