The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001828.5 | NP_001819.2 | 2 - 3 | 172 | 74 |
GenBank mRNA | AV715261.1 | - | 2 - 3 | 171 | 74 |
BC119711.1 | - | 2 - 3 | 74 | ||
BC119712.1 | - | 2 - 3 | 74 | ||
FJ613334.1 | - | 2 - 3 | 115 | 74 | |
FJ613335.1 | - | 2 - 3 | 74 | ||
L01664.1 | - | 2 - 3 | 74 |