The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001256524.1 | NP_001243453.1 | 3 - 4 | 406 | 88 |
NM_001256525.1 | NP_001243454.1 | 2 - 3 | 306 | 88 | |
NM_003426.3 | NP_003417.2 | 2 - 3 | 634 | 88 | |
GenBank mRNA | AK291358.1 | - | 3 - 4 | 332 | 88 |
AK299569.1 | - | 2 - 3 | 232 | 88 | |
BC013395.2 | - | 2 - 3 | 232 | 88 | |
BC048295.1 | - | 2 - 3 | 219 | 88 | |
CR456616.1 | - | 2 - 3 | 173 | 88 | |
X71623.1 | - | 1 - 2 | 69 | 88 | |
X92715.1 | - | 1 - 2 | 1235 | 88 |