The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001256339.1 | NP_001243268.1 | 2 - 3 | 66 | 78 |
NM_006167.3 | NP_006158.2 | 1 - 2 | 339 | 78 | |
GenBank mRNA | AF247704.1 | - | 1 - 2 | 339 | 78 |
AF249669.1 | - | 2 - 3 | 169 | 78 | |
AF249670.1 | - | 1 - 2 | 154 | 78 | |
AF249671.1 | - | 2 - 3 | 94 | 78 | |
AF249672.1 | - | 2 - 3 | 76 | 78 | |
BC074863.2 | - | 1 - 2 | 323 | 78 | |
BC074864.2 | - | 1 - 2 | 323 | 78 | |
U80669.1 | - | 1 - 2 | 320 | 78 | |
U91540.1 | - | 1 - 2 | 291 | 78 |