The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000553.4 | NP_000544.2 | 32 - 33 | 4608 | 85 |
XM_011544639.2 | XP_011542941.1 | 31 - 32 | 4009 | 85 | |
XM_011544640.1 | XP_011542942.1 | 20 - 21 | 2321 | 85 | |
GenBank mRNA | AB209652.1 | - | 18 - 19 | 2051 | 85 |
AF091214.1 | - | 32 - 33 | 4051 | 85 | |
AL833572.1 | - | 20 - 21 | 2454 | 85 | |
AY818673.1 | - | 31 - 32 | 3820 | 85 |