The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001220765.2 | NP_001207694.1 | 3 - 4 | 429 | 76 |
NM_001220768.2 | NP_001207697.1 | 2 - 3 | 175 | 76 | |
NM_001220771.2 | NP_001207700.1 | 2 - 3 | 175 | 76 | |
NM_001291837.1 | NP_001278766.1 | 3 - 4 | 402 | 76 | |
NM_006060.5 | NP_006051.1 | 3 - 4 | 429 | 76 | |
XM_011515061.2 | XP_011513363.1 | 3 - 4 | 257 | 76 | |
XM_011515068.2 | XP_011513370.1 | 3 - 4 | 456 | 76 | |
XM_011515069.2 | XP_011513371.1 | 3 - 4 | 4084 | 76 | |
XM_011515070.2 | XP_011513372.1 | 3 - 4 | 260 | 76 | |
XM_017011669.1 | XP_016867158.1 | 3 - 4 | 224 | 76 | |
XM_017011670.1 | XP_016867159.1 | 3 - 4 | 585 | 76 | |
GenBank mRNA | AK308411.1 | - | 3 - 4 | 359 | 76 |
AY377974.1 | - | 3 - 4 | 329 | 76 | |
BC018349.1 | - | 3 - 4 | 353 | 76 | |
BT009836.1 | - | 2 - 3 | 161 | 76 | |
DB122945.1 | - | 3 - 4 | 402 | 76 | |
S80876.1 | - | 1 - 2 | 398 | 76 | |
U40462.1 | - | 3 - 4 | 329 | 76 |