The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000905.3 | NP_000896.1 | 3 - 4 | 361 | 70 |
GenBank mRNA | AI198311.1 | - | 1 - 2 | 70 | |
BC029497.1 | - | 3 - 4 | 361 | 70 | |
K01911.1 | - | 3 - 4 | 359 | 70 | |
M15789.1 | - | 2 - 3 | 287 | 70 |